siRNA Details
Gene Symbol: FAM183BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028347.1 | 2 | 1141 | |||
GenBank mRNA | AK124132.1 | - | 1 | 1238 | ||
BC045803.1 | - | 2 | 1089 | |||
FJ211598.1 | - | 1 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
