siRNA Details
Gene Symbol: SMAD5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026763.1 | 2 | 1591 | |||
GenBank mRNA | AF071111.1 | - | 2 | 1591 | ||
AF086556.1 | - | 2 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
