siRNA Details
Gene Symbol: UMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302348.1 | NP_001289277.1 | 4 | 621 | ||
NM_001302349.1 | NP_001289278.1 | 4 | 614 | |||
NM_001302350.1 | NP_001289279.1 | 5 | 728 | |||
GenBank mRNA | AK027125.1 | - | 2 | 280 | ||
AK295253.1 | - | 5 | 482 | |||
DC313244.1 | - | 4 | 466 | |||
DC354992.1 | - | 4 | 516 | |||
DC368552.1 | - | 4 | 505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
