siRNA Details
Gene Symbol: SYTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009991.3 | NP_001009991.2 | 16 | 1828 | ||
NM_001242384.1 | NP_001229313.1 | 18 | 2048 | |||
NM_001242394.1 | NP_001229323.1 | 17 | 2440 | |||
NM_001242395.1 | NP_001229324.1 | 15 | 2236 | |||
NM_001318745.1 | NP_001305674.1 | 14 | 1762 | |||
XM_005267215.4 | XP_005267272.1 | 17 | 2538 | |||
XM_005267218.4 | XP_005267275.1 | 17 | 1849 | |||
XM_005267222.4 | XP_005267279.1 | 9 | 1120 | |||
XM_006715605.3 | XP_006715668.1 | 18 | 2630 | |||
XM_006715606.3 | XP_006715669.1 | 18 | 2551 | |||
XM_006715611.3 | XP_006715674.1 | 17 | 2153 | |||
XM_011536254.2 | XP_011534556.1 | 18 | 2319 | |||
XM_011536255.2 | XP_011534557.1 | 8 | 1375 | |||
XM_017011496.1 | XP_016866985.1 | 17 | 2719 | |||
XM_017011497.1 | XP_016866986.1 | 18 | 2056 | |||
XM_017011498.1 | XP_016866987.1 | 14 | 1869 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
