siRNA Details
Gene Symbol: SYTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009991.3 | NP_001009991.2 | 14 | 1530 | ||
NM_001242384.1 | NP_001229313.1 | 16 | 1750 | |||
NM_001242394.1 | NP_001229323.1 | 15 | 2142 | |||
NM_001242395.1 | NP_001229324.1 | 13 | 1938 | |||
NM_001318745.1 | NP_001305674.1 | 12 | 1464 | |||
XM_005267215.4 | XP_005267272.1 | 15 | 2240 | |||
XM_005267218.4 | XP_005267275.1 | 15 | 1551 | |||
XM_005267222.4 | XP_005267279.1 | 7 | 822 | |||
XM_006715605.3 | XP_006715668.1 | 16 | 2332 | |||
XM_006715606.3 | XP_006715669.1 | 16 | 2253 | |||
XM_006715611.3 | XP_006715674.1 | 15 | 1855 | |||
XM_011536254.2 | XP_011534556.1 | 16 | 2021 | |||
XM_011536255.2 | XP_011534557.1 | 6 | 1077 | |||
XM_017011495.1 | XP_016866984.1 | 13 | 2209 | |||
XM_017011496.1 | XP_016866985.1 | 15 | 2421 | |||
XM_017011497.1 | XP_016866986.1 | 16 | 1758 | |||
XM_017011498.1 | XP_016866987.1 | 12 | 1571 | |||
GenBank mRNA | AK131092.1 | - | 11 | 3893 | ||
AK298712.1 | - | 16 | 1710 | |||
AK304191.1 | - | 7 | 1007 | |||
BC085612.1 | - | 13 | 1476 | 30707698 | ||
BC100830.1 | - | 14 | 1531 | 40001431 | ||
BC100831.1 | - | 14 | 1531 | 40001432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
