siRNA Details
Gene Symbol: XKR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543821.2 | XP_011542123.1 | 2 | 852 | ||
GenBank mRNA | AJ301560.1 | - | 2 | 782 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
