siRNA Details
Gene Symbol: INTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080453.2 | NP_001073922.2 | 3 | 413 | ||
XM_011515260.1 | XP_011513562.1 | 3 | 413 | |||
XM_011515262.2 | XP_011513564.1 | 3 | 413 | |||
XM_017011959.1 | XP_016867448.1 | 3 | 413 | |||
XM_017011960.1 | XP_016867449.1 | 3 | 413 | |||
GenBank mRNA | AI935093.1 | - | 1 | 134 | ||
DB113520.1 | - | 3 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
