siRNA Details
Gene Symbol: INTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080453.2 | NP_001073922.2 | 44, 45 | 6295 | ||
XM_011515260.1 | XP_011513562.1 | 44, 45 | 6325 | |||
XM_017011959.1 | XP_016867448.1 | 44, 45 | 6325 | |||
XM_017011960.1 | XP_016867449.1 | 44, 45 | 6295 | |||
GenBank mRNA | AB037861.1 | - | 26, 27 | 3755 | ||
AL050110.1 | - | 11, 12 | 1545 | |||
AL137358.1 | - | 4, 5 | 447 | |||
AY358482.1 | - | 19, 20 | 2756 | |||
BC004286.1 | - | 25, 26 | 3468 | |||
BC013367.2 | - | 14, 15 | 2051 | |||
BC018777.1 | - | 12, 13 | 1718 | |||
BC069262.1 | - | 16, 17 | 2325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
