siRNA Details
Gene Symbol: INTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080453.2 | NP_001073922.2 | 45 | 6312 | ||
XM_011515260.1 | XP_011513562.1 | 45 | 6342 | |||
XM_017011959.1 | XP_016867448.1 | 45 | 6342 | |||
XM_017011960.1 | XP_016867449.1 | 45 | 6312 | |||
GenBank mRNA | AB037861.1 | - | 27 | 3772 | ||
AL050110.1 | - | 12 | 1562 | |||
AL137358.1 | - | 5 | 464 | |||
AY358482.1 | - | 20 | 2773 | |||
BC004286.1 | - | 26 | 3485 | |||
BC013367.2 | - | 15 | 2068 | |||
BC018777.1 | - | 13 | 1735 | |||
BC069262.1 | - | 17 | 2342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
