siRNA Details
Gene Symbol: GBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098834.2 | NP_001092304.1 | 2 | 702 | ||
XM_017011964.1 | XP_016867453.1 | 3 | 719 | |||
XM_017011965.1 | XP_016867454.1 | 3 | 716 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
