siRNA Details
Gene Symbol: FER1L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039112.2 | NP_001034201.2 | 10 | 1203 | ||
XM_006716618.3 | XP_006716681.1 | 10 | 1130 | |||
XM_011517231.2 | XP_011515533.1 | 11 | 1169 | |||
XM_011517232.2 | XP_011515534.1 | 11 | 1310 | |||
XM_011517233.2 | XP_011515535.1 | 10 | 1271 | |||
XM_011517235.2 | XP_011515537.1 | 11 | 1170 | |||
GenBank mRNA | AB196633.1 | - | 10 | 1203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
