siRNA Details
Gene Symbol: SUGT1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003667.3 | 4, 5 | 711 | |||
GenBank mRNA | DA804368.1 | - | 4, 5 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
