siRNA Details
Gene Symbol: KDM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153042.3 | NP_694587.3 | 11 | 1166 | ||
XM_005248925.1 | XP_005248982.1 | 13 | 1562 | |||
XM_005248926.1 | XP_005248983.1 | 13 | 1562 | |||
XM_005248928.1 | XP_005248985.1 | 10 | 1127 | |||
XM_011514385.1 | XP_011512687.1 | 11 | 1166 | |||
XM_011514386.1 | XP_011512688.1 | 11 | 1166 | |||
XM_017010440.1 | XP_016865929.1 | 14 | 1589 | |||
XM_017010441.1 | XP_016865930.1 | 14 | 1589 | |||
XM_017010442.1 | XP_016865931.1 | 13 | 1435 | |||
XM_017010443.1 | XP_016865932.1 | 13 | 1382 | |||
XM_017010445.1 | XP_016865934.1 | 12 | 1193 | |||
XM_017010446.1 | XP_016865935.1 | 14 | 1589 | |||
GenBank mRNA | AK125318.1 | - | 11 | 1102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
