siRNA Details
Gene Symbol: KDM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153042.3 | NP_694587.3 | 17 | 1884 | ||
XM_005248925.1 | XP_005248982.1 | 21 | 2583 | |||
XM_005248926.1 | XP_005248983.1 | 21 | 2580 | |||
XM_005248928.1 | XP_005248985.1 | 18 | 2148 | |||
XM_011514385.1 | XP_011512687.1 | 19 | 2187 | |||
XM_011514386.1 | XP_011512688.1 | 19 | 2184 | |||
XM_017010440.1 | XP_016865929.1 | 22 | 2610 | |||
XM_017010441.1 | XP_016865930.1 | 22 | 2607 | |||
XM_017010442.1 | XP_016865931.1 | 21 | 2456 | |||
XM_017010443.1 | XP_016865932.1 | 21 | 2403 | |||
XM_017010445.1 | XP_016865934.1 | 20 | 2214 | |||
GenBank mRNA | AK091217.1 | - | 4 | 556 | ||
AK125318.1 | - | 17 | 1820 | |||
BC113093.1 | - | 4 | 315 | |||
CR627410.1 | - | 4 | 1467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
