siRNA Details
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 18 | 2596 | ||
XM_005273409.1 | XP_005273466.1 | 19 | 2707 | |||
XM_005273410.1 | XP_005273467.1 | 19 | 2683 | |||
XM_005273411.1 | XP_005273468.1 | 17 | 2515 | |||
XM_017013088.1 | XP_016868577.1 | 16 | 2404 | |||
XM_017013089.1 | XP_016868578.1 | 17 | 2469 | |||
GenBank mRNA | AK057293.1 | - | 9 | 1986 | ||
AK074839.1 | - | 5 | 1166 | |||
AL832870.1 | - | 1 | 145 | |||
CR749838.1 | - | 14 | 2020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
