siRNA Details
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 18 | 3408 | ||
XM_005273409.1 | XP_005273466.1 | 19 | 3519 | |||
XM_005273410.1 | XP_005273467.1 | 19 | 3495 | |||
XM_005273411.1 | XP_005273468.1 | 17 | 3327 | |||
XM_017013088.1 | XP_016868577.1 | 16 | 3216 | |||
XM_017013089.1 | XP_016868578.1 | 17 | 3281 | |||
GenBank mRNA | AK025895.1 | - | 1 | 481 | ||
AK074839.1 | - | 5 | 1978 | |||
AL832870.1 | - | 1 | 957 | |||
BC012452.1 | - | 1 | 431 | |||
CR457352.1 | - | 1 | 231 | |||
CR749838.1 | - | 14 | 2832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
