siRNA Details
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152419.2 | NP_689632.2 | 18 | 2294 | ||
XM_005273409.1 | XP_005273466.1 | 19 | 2405 | |||
XM_005273410.1 | XP_005273467.1 | 19 | 2381 | |||
XM_005273411.1 | XP_005273468.1 | 17 | 2213 | |||
XM_017013088.1 | XP_016868577.1 | 16 | 2102 | |||
XM_017013089.1 | XP_016868578.1 | 17 | 2167 | |||
GenBank mRNA | AK057293.1 | - | 9 | 1684 | ||
AK304441.1 | - | 18 | 2246 | |||
CR749838.1 | - | 14 | 1718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
