siRNA Details
Gene Symbol: KCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135914.1 | NP_001129386.1 | 31, 32 | 3608 | ||
XM_017012184.1 | XP_016867673.1 | 35, 36 | 3998 | |||
XM_017012185.1 | XP_016867674.1 | 35, 36 | 3983 | |||
XM_017012186.1 | XP_016867675.1 | 35, 36 | 3998 | |||
XM_017012187.1 | XP_016867676.1 | 35, 36 | 3998 | |||
XM_017012188.1 | XP_016867677.1 | 35, 36 | 3998 | |||
XM_017012189.1 | XP_016867678.1 | 35, 36 | 3998 | |||
XM_017012190.1 | XP_016867679.1 | 35, 36 | 3998 | |||
XM_017012191.1 | XP_016867680.1 | 28, 29 | 3217 | |||
XM_017012192.1 | XP_016867681.1 | 28, 29 | 3228 | |||
GenBank mRNA | AK090684.1 | - | 3, 4 | 629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
