siRNA Details
Gene Symbol: SVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153366.3 | NP_699197.3 | 37 | 6967 | ||
GenBank mRNA | AJ619977.1 | - | 37 | 6802 | ||
AY243503.1 | - | 37 | 6802 | |||
AY916667.1 | - | 1 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
