siRNA Details
Gene Symbol: SVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153366.3 | NP_699197.3 | 38 | 9751 | ||
GenBank mRNA | AJ619977.1 | - | 38 | 9586 | ||
AK075200.1 | - | 1 | 206 | |||
AK075234.1 | - | 1 | 2429 | |||
AL832416.1 | - | 1 | 2238 | |||
AY243503.1 | - | 38 | 9586 | |||
AY916667.1 | - | 2 | 3192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
