siRNA Details
Gene Symbol: FAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008781.2 | NP_001008781.2 | 18 | 11187 | ||
XM_017017178.1 | XP_016872667.1 | 20 | 11372 | |||
XM_017017179.1 | XP_016872668.1 | 21 | 11594 | |||
XM_017017180.1 | XP_016872669.1 | 20 | 11371 | |||
XM_017017181.1 | XP_016872670.1 | 19 | 11302 | |||
XM_017017182.1 | XP_016872671.1 | 20 | 11371 | |||
XM_017017183.1 | XP_016872672.1 | 19 | 11302 | |||
XM_017017184.1 | XP_016872673.1 | 20 | 11370 | |||
XM_017017185.1 | XP_016872674.1 | 19 | 9198 | |||
XM_017017186.1 | XP_016872675.1 | 19 | 8486 | |||
XM_017017187.1 | XP_016872676.1 | 19 | 12844 | |||
GenBank mRNA | AB076400.1 | - | 2 | 695 | ||
AK126620.1 | - | 1 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
