siRNA Details
Gene Symbol: FAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008781.2 | NP_001008781.2 | 25 | 13919 | ||
XM_017017178.1 | XP_016872667.1 | 29 | 14200 | |||
XM_017017179.1 | XP_016872668.1 | 30 | 14422 | |||
XM_017017180.1 | XP_016872669.1 | 28 | 14139 | |||
XM_017017181.1 | XP_016872670.1 | 28 | 14130 | |||
XM_017017182.1 | XP_016872671.1 | 27 | 14103 | |||
XM_017017183.1 | XP_016872672.1 | 26 | 14034 | |||
XM_017017185.1 | XP_016872674.1 | 28 | 12026 | |||
XM_017017186.1 | XP_016872675.1 | 28 | 11314 | |||
XM_017017187.1 | XP_016872676.1 | 28 | 15672 | |||
GenBank mRNA | AB076400.1 | - | 11 | 3523 | ||
AK126620.1 | - | 8 | 2930 | |||
AK127104.1 | - | 3 | 1060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
