siRNA Details
Gene Symbol: NUTM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278495.1 | NP_001265424.1 | 7 | 2580 |
Gene Symbol: NUTM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099338.1 | NP_001092808.1 | 7 | 2580 | ||
XM_011540122.2 | XP_011538424.1 | 9 | 5423 | |||
GenBank mRNA | BC092519.1 | - | 7 | 2576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
