siRNA Details
Gene Symbol: C10orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039380.3 | NP_001034469.2 | 2 | 1456 | ||
NR_126421.1 | 3 | 593 | ||||
XM_017015895.1 | XP_016871384.1 | 3 | 587 | |||
GenBank mRNA | AK055129.1 | - | 2 | 1434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
