siRNA Details
Gene Symbol: DRGX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276451.1 | NP_001263380.1 | 4 | 363 | ||
XM_011540089.2 | XP_011538391.1 | 3 | 281 | |||
GenBank mRNA | HY131276.1 | - | 4 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
