siRNA Details
Gene Symbol: TAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031923.3 | NP_114129.1 | 3 | 1972 | ||
XM_011519741.1 | XP_011518043.1 | 3 | 1996 | |||
GenBank mRNA | AJ292190.1 | - | 3 | 1766 | ||
AK307521.1 | - | 3 | 1945 | |||
AL117661.1 | - | 3 | 1700 | |||
BC028077.1 | - | 3 | 1929 | |||
BC062352.1 | - | 3 | 1939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
