siRNA Details
Gene Symbol: SOHLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012415.2 | NP_001012415.2 | 7 | 1916 | ||
NM_001101677.1 | NP_001095147.1 | 8 | 1344 | |||
XM_005263403.3 | XP_005263460.1 | 9 | 1361 | |||
XM_006717109.3 | XP_006717172.1 | 10 | 2486 | |||
XM_011518698.2 | XP_011517000.1 | 11 | 2515 | |||
GenBank mRNA | AY902244.1 | - | 7 | 1916 | ||
BC031861.1 | - | 7 | 1927 | 4826967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
