siRNA Details
Gene Symbol: KIAA1462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020848.2 | NP_065899.1 | 3 | 2640 | ||
XM_011519608.1 | XP_011517910.1 | 3 | 2679 | |||
XM_011519609.2 | XP_011517911.1 | 4 | 2707 | |||
GenBank mRNA | AB040895.2 | - | 2 | 2533 | ||
AK308850.1 | - | 3 | 2622 | |||
AL583916.1 | - | 1 | 577 | |||
BX648192.1 | - | 4 | 2729 | |||
BX648263.1 | - | 1 | 1564 | |||
BX648773.1 | - | 1 | 1690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
