siRNA Details
Gene Symbol: KIAA1462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020848.2 | NP_065899.1 | 3 | 3145 | ||
XM_011519608.1 | XP_011517910.1 | 3 | 3184 | |||
XM_011519609.2 | XP_011517911.1 | 4 | 3212 | |||
GenBank mRNA | AB040895.2 | - | 2 | 3038 | ||
AL583916.1 | - | 1 | 1082 | |||
BC047548.1 | - | 1 | 458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
