siRNA Details
Gene Symbol: ERICH6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182542.2 | NP_872348.2 | 15 | 2342 | ||
XM_011534965.2 | XP_011533267.1 | 14 | 2361 | |||
XM_017020418.1 | XP_016875907.1 | 14 | 2289 | |||
GenBank mRNA | AK057244.1 | - | 15 | 2324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
