siRNA Details
Gene Symbol: HSPA12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025015.2 | NP_079291.2 | 6 | 701 | ||
XM_005269672.3 | XP_005269729.1 | 7 | 851 | |||
XM_005269673.4 | XP_005269730.1 | 7 | 1795 | |||
XM_011539579.2 | XP_011537881.1 | 8 | 912 | |||
XM_011539580.2 | XP_011537882.1 | 7 | 857 | |||
XM_017016032.1 | XP_016871521.1 | 4 | 423 | |||
GenBank mRNA | AB007877.1 | - | 6 | 701 | ||
AK023936.1 | - | 1 | 736 | |||
AK294672.1 | - | 5 | 515 | |||
AK294884.1 | - | 1 | 20 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
