siRNA Details
Gene Symbol: HSPA12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025015.2 | NP_079291.2 | 12 | 3161 | ||
XM_005269672.3 | XP_005269729.1 | 13 | 3311 | |||
XM_005269673.4 | XP_005269730.1 | 13 | 4255 | |||
XM_011539579.2 | XP_011537881.1 | 14 | 3372 | |||
XM_011539580.2 | XP_011537882.1 | 13 | 3317 | |||
XM_017016032.1 | XP_016871521.1 | 10 | 2883 | |||
GenBank mRNA | AB007877.1 | - | 12 | 3161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
