siRNA Details
Gene Symbol: ARHGAP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152432.2 | NP_689645.2 | 21 | 2257 | ||
XM_011542615.2 | XP_011540917.1 | 21 | 4745 | |||
XM_011542616.2 | XP_011540918.1 | 21 | 2514 | |||
XM_011542617.2 | XP_011540919.1 | 22 | 2400 | |||
XM_017017237.1 | XP_016872726.1 | 21 | 2703 | |||
XM_017017238.1 | XP_016872727.1 | 22 | 4886 | |||
GenBank mRNA | AK023666.1 | - | 2 | 313 | ||
AK057372.1 | - | 9 | 1127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
