siRNA Details
Gene Symbol: ARHGAP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152432.2 | NP_689645.2 | 7 | 678 | ||
XM_011542615.2 | XP_011540917.1 | 7 | 3166 | |||
XM_011542616.2 | XP_011540918.1 | 7 | 935 | |||
XM_011542617.2 | XP_011540919.1 | 8 | 821 | |||
XM_017017237.1 | XP_016872726.1 | 7 | 1124 | |||
XM_017017238.1 | XP_016872727.1 | 8 | 3307 | |||
GenBank mRNA | DA800121.1 | - | 3 | 243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
