siRNA Details
Gene Symbol: SMCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145010.1 | NP_001138482.1 | 2 | 195 | ||
XM_011520634.2 | XP_011518936.1 | 2 | 373 | |||
XM_011520636.2 | XP_011518938.1 | 2 | 373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
