siRNA Details
Gene Symbol: C1QL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010908.1 | NP_001010908.1 | 1, 2 | 1528 | ||
GenBank mRNA | BC127716.1 | - | 2, 3 | 596 | 40127920 | |
BC127717.1 | - | 2, 3 | 542 | 40127921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
