siRNA Details
Gene Symbol: C1QL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010908.1 | NP_001010908.1 | 1, 2 | 1530 | ||
GenBank mRNA | BC127716.1 | - | 2, 3 | 598 | 40127920 | |
BC127717.1 | - | 2, 3 | 544 | 40127921 |

More Information
Other Species Targeted

Gene Ontology Categories:
