siRNA Details
Gene Symbol: LRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039029.2 | NP_001034118.1 | 5 | 3859 | ||
NM_001163925.1 | NP_001157397.1 | 5 | 3895 | |||
NM_001163926.1 | NP_001157398.1 | 5 | 3862 | |||
XM_011521015.1 | XP_011519317.1 | 5 | 4080 | |||
XM_017019848.1 | XP_016875337.1 | 3 | 3267 | |||
GenBank mRNA | AK095610.1 | - | 5 | 3597 | ||
AK124687.1 | - | 5 | 3466 | |||
AK125402.1 | - | 2 | 2683 | |||
AK126866.1 | - | 1 | 3887 | |||
BC036868.1 | - | 1 | 2316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
