siRNA Details
Gene Symbol: LRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039029.2 | NP_001034118.1 | 4 | 1044 | ||
NM_001163925.1 | NP_001157397.1 | 4 | 1080 | |||
NM_001163926.1 | NP_001157398.1 | 4 | 1047 | |||
XM_011521015.1 | XP_011519317.1 | 4 | 1265 | |||
XM_017019848.1 | XP_016875337.1 | 2 | 452 | |||
GenBank mRNA | AK095610.1 | - | 4 | 782 | ||
AK124687.1 | - | 4 | 651 | |||
BC146894.1 | - | 4 | 683 | 9056822 | ||
BC146910.1 | - | 4 | 683 | 9056838 | ||
BC150568.1 | - | 2 | 288 | 40127648 | ||
DA418133.1 | - | 2 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
