siRNA Details
Gene Symbol: ASPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080464.2 | NP_001073933.2 | 3 | 299 | ||
XM_005267590.4 | XP_005267647.1 | 3 | 325 | |||
XM_017021265.1 | XP_016876754.1 | 1 | 766 | |||
XM_017021266.1 | XP_016876755.1 | 1 | 766 | |||
XM_017021267.1 | XP_016876756.1 | 1 | 766 | |||
XM_017021268.1 | XP_016876757.1 | 3 | 327 | |||
XM_017021269.1 | XP_016876758.1 | 1 | 766 | |||
XM_017021270.1 | XP_016876759.1 | 1 | 766 | |||
XM_017021271.1 | XP_016876760.1 | 1 | 766 | |||
XM_017021272.1 | XP_016876761.1 | 1 | 766 | |||
XM_017021273.1 | XP_016876762.1 | 1 | 766 | |||
XM_017021274.1 | XP_016876763.1 | 3 | 231 | |||
XM_017021275.1 | XP_016876764.1 | 1 | 766 | |||
XM_017021276.1 | XP_016876765.1 | 1 | 766 | |||
GenBank mRNA | BC035836.1 | - | 3 | 274 | ||
BC136637.1 | - | 3 | 241 | 9020627 | ||
BX247999.1 | - | 2 | 1048 | |||
BX280385.1 | - | 3 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
