siRNA Details
Gene Symbol: CCDC182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282544.1 | NP_001269473.1 | 1 | 128 | ||
GenBank mRNA | EG357680.1 | - | 1 | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
