siRNA Details
Gene Symbol: MACROD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033087.1 | NP_001028259.1 | 2 | 260 | ||
NM_080676.5 | NP_542407.2 | 6 | 767 | |||
XM_017027677.1 | XP_016883166.1 | 6 | 791 | |||
GenBank mRNA | AK131348.1 | - | 6 | 897 | ||
BC018687.1 | - | 2 | 263 | 4717477 | ||
DR001087.1 | - | 326 | ||||
EF159161.1 | - | 503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
