siRNA Details
Gene Symbol: ZMAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011657.3 | NP_001011657.2 | 6 | 873 | ||
NM_001282400.1 | NP_001269329.1 | 9 | 1429 | |||
NM_001282401.1 | NP_001269330.1 | 8 | 1376 | |||
XM_005262212.4 | XP_005262269.2 | 6 | 890 | |||
XM_005262213.4 | XP_005262270.2 | 6 | 887 | |||
XM_005262214.3 | XP_005262271.2 | 5 | 821 | |||
XM_005262216.4 | XP_005262273.2 | 6 | 592 | |||
XM_006724711.3 | XP_006724774.1 | 2 | 352 | |||
XM_017029903.1 | XP_016885392.1 | 6 | 589 | |||
XM_017029904.1 | XP_016885393.1 | 6 | 2642 | |||
GenBank mRNA | AB058692.1 | - | 1 | 3506 | ||
AK094141.1 | - | 9 | 1390 | |||
AK296520.1 | - | 8 | 1341 | |||
BC140920.1 | - | 1 | 3472 | 9056732 | ||
BC152470.1 | - | 1 | 3506 | 8862607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
