siRNA Details
Gene Symbol: RNF169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098638.1 | NP_001092108.1 | 6 | 3483 | ||
XM_011544889.2 | XP_011543191.1 | 6 | 3558 | |||
GenBank mRNA | AB082522.1 | - | 6 | 3483 | ||
BX640750.1 | - | 1 | 2387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
