siRNA Details
Gene Symbol: PRTG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173814.5 | NP_776175.2 | 13 | 2466 | ||
XM_017022081.1 | XP_016877570.1 | 13 | 2553 | |||
XM_017022082.1 | XP_016877571.1 | 13 | 2996 | |||
GenBank mRNA | AK098622.1 | - | 4 | 474 | ||
AY630258.1 | - | 13 | 2326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
