siRNA Details
Gene Symbol: TMEM41B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015012.3 | NP_055827.1 | 7 | 1083 | ||
NR_028491.2 | 7 | 1083 | ||||
GenBank mRNA | AK303339.1 | - | 7 | 774 | ||
BC035034.1 | - | 7 | 961 | 5259179 | ||
BC044597.1 | - | 7 | 952 | 4811565 | ||
BC064366.1 | - | 7 | 840 | |||
BC091524.1 | - | 7 | 882 | 30512350 | ||
D26067.1 | - | 7 | 889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
