siRNA Details
Gene Symbol: BEAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136106.4 | NP_001129578.1 | 4 | 1009 | ||
NM_001178020.2 | NP_001171491.1 | 5 | 1116 | |||
XM_011522883.1 | XP_011521185.1 | 5 | 1222 | |||
XM_011522884.1 | XP_011521186.1 | 5 | 1527 | |||
XM_011522885.2 | XP_011521187.1 | 3 | 1369 | |||
XM_011522888.1 | XP_011521190.1 | 6 | 1723 | |||
XM_011522889.1 | XP_011521191.1 | 5 | 1538 | |||
XM_011522890.1 | XP_011521192.1 | 6 | 2028 | |||
XM_011522891.1 | XP_011521193.1 | 6 | 1407 | |||
XM_011522892.2 | XP_011521194.1 | 4 | 1676 | |||
GenBank mRNA | AB472390.1 | - | 5 | 1042 | ||
AB472391.1 | - | 4 | 935 | |||
AK056142.1 | - | 4 | 969 | |||
BC107477.1 | - | 3 | 871 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
