siRNA Details
Gene Symbol: BEAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136106.4 | NP_001129578.1 | 4 | 2659 | ||
NM_001178020.2 | NP_001171491.1 | 5 | 2766 | |||
XM_011522883.1 | XP_011521185.1 | 5 | 2872 | |||
XM_011522884.1 | XP_011521186.1 | 5 | 3177 | |||
XM_011522885.2 | XP_011521187.1 | 3 | 3019 | |||
XM_011522888.1 | XP_011521190.1 | 6 | 3373 | |||
XM_011522889.1 | XP_011521191.1 | 5 | 3188 | |||
XM_011522890.1 | XP_011521192.1 | 6 | 3678 | |||
XM_011522891.1 | XP_011521193.1 | 6 | 3057 | |||
XM_011522892.2 | XP_011521194.1 | 4 | 3326 | |||
GenBank mRNA | AF070610.1 | - | 1 | 1465 | ||
AK056142.1 | - | 5 | 2321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
