siRNA Details
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243965.1 | NP_001230894.1 | 2 | 216 | ||
GenBank mRNA | AF017441.1 | - | 1 | 35 | ||
AJ297583.1 | - | 2 | 122 | |||
AJ297587.1 | - | 2 | 122 | |||
AJ697892.1 | - | 2 | 131 | |||
AJ697893.1 | - | 2 | 131 | |||
AY961065.1 | - | 2 | 122 | |||
AY961066.1 | - | 2 | 122 | |||
BC007920.2 | - | 2 | 168 | 4309837 | ||
BC008403.1 | - | 2 | 185 | 4272571 | ||
BC031023.1 | - | 2 | 185 | 4695321 | ||
DB246210.1 | - | 2 | 226 | |||
L76133.1 | - | 2 | 162 | |||
M11867.1 | - | 2 | 151 | |||
M25265.1 | - | 1 | 35 | |||
M25266.1 | - | 1 | 35 | |||
U37582.1 | - | 1 | 35 | |||
U65585.1 | - | 2 | 162 | |||
U95817.1 | - | 1 | 35 | |||
X00699.1 | - | 2 | 157 |
Gene Symbol: HLA-DRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225683.1 | - | 2 | 185 | ||
AK225690.1 | - | 2 | 185 | |||
AK291499.1 | - | 2 | 185 | |||
AK293020.1 | - | 2 | 226 | |||
AY961064.1 | - | 2 | 122 | |||
AY961067.1 | - | 2 | 122 | |||
AY961070.2 | - | 2 | 122 | |||
M59803.1 | - | 1 | 35 | |||
U17379.1 | - | 1 | 49 | |||
U29342.1 | - | 1 | 35 | |||
U35375.1 | - | 1 | 35 | |||
U35729.1 | - | 1 | 35 | |||
U37433.1 | - | 1 | 35 | |||
U47028.1 | - | 1 | 35 |
Gene Symbol: HLA-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB062112.2 | - | 2 | 166 | ||
AF490771.1 | - | 2 | 103 | |||
AJ297582.1 | - | 2 | 122 | |||
AJ580838.1 | - | 1 | 22 | |||
M17379.1 | - | 1 | 35 | |||
M17382.1 | - | 1 | 35 | |||
M17383.1 | - | 1 | 35 | |||
M27689.1 | - | 1 | 35 | |||
M59798.1 | - | 1 | 35 | |||
U17381.1 | - | 2 | 49 | |||
U95818.1 | - | 14 | ||||
X99895.1 | - | 2 | 129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
