siRNA Details
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243965.1 | NP_001230894.1 | 3 | 522 | ||
GenBank mRNA | U65585.1 | - | 3 | 468 | ||
X00699.1 | - | 3 | 463 |
Gene Symbol: HLA-DRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK291499.1 | - | 3 | 491 | ||
AY961064.1 | - | 3 | 428 | |||
AY961067.1 | - | 3 | 428 | |||
AY961070.2 | - | 3 | 428 |
Gene Symbol: HLA-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M17379.1 | - | 2 | 341 | ||
X99895.1 | - | 3 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
